siRNA Details
| Gene Symbol: Scgb3a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054037.2 | NP_473378.1 | 506 | |||
| NM_170727.2 | NP_733923.1 | 611 | ||||
| GenBank mRNA | AF313456.1 | - | 491 | |||
| AF313457.1 | - | 596 | ||||
| BC086910.1 | - | 488 | 6759656 | |||

Scgb3a1
Genomic Map