siRNA Details
| Gene Symbol: Erdr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133362.2 | NP_579940.1 | 577 | |||
| GenBank mRNA | AK051407.1 | - | 1077 | |||
| BC018296.1 | - | 582 | 3499536 | |||
| BC038133.1 | - | 391 | ||||
| BC058113.1 | - | 2 | 423 | 6820436 | ||
| BC080795.1 | - | 432 | 5006491 | |||
| BC082282.1 | - | 529 | ||||
| BC086616.1 | - | 2833 | ||||

Erdr1
Genomic Map