siRNA Details
| Gene Symbol: Vax1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009501.1 | NP_033527.1 | 2 | 416 | ||
| GenBank mRNA | AF064554.1 | - | 2 | 416 | ||
| BC111818.1 | - | 2 | 415 | 40044698 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vax1| Gene Symbol: Vax1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009501.1 | NP_033527.1 | 2 | 416 | ||
| GenBank mRNA | AF064554.1 | - | 2 | 416 | ||
| BC111818.1 | - | 2 | 415 | 40044698 | ||