siRNA Details
| Gene Symbol: Ptcra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | U16958.1 | - | 4 | 1063 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptcra| Gene Symbol: Ptcra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | U16958.1 | - | 4 | 1063 | ||