siRNA Details
| Gene Symbol: Sprr2k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011477.3 | NP_035607.2 | 2 | 244 | ||
| GenBank mRNA | AY158995.1 | - | 2 | 244 | ||
| BC116866.1 | - | 2 | 230 | |||
| BC116868.1 | - | 2 | 230 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sprr2k| Gene Symbol: Sprr2k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011477.3 | NP_035607.2 | 2 | 244 | ||
| GenBank mRNA | AY158995.1 | - | 2 | 244 | ||
| BC116866.1 | - | 2 | 230 | |||
| BC116868.1 | - | 2 | 230 | |||