siRNA Details
| Gene Symbol: Rgs9bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145840.3 | NP_665839.1 | 1 | 847 | ||
| GenBank mRNA | AF483907.1 | - | 1 | 485 | ||
| AY229888.1 | - | 1 | 809 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rgs9bp| Gene Symbol: Rgs9bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145840.3 | NP_665839.1 | 1 | 847 | ||
| GenBank mRNA | AF483907.1 | - | 1 | 485 | ||
| AY229888.1 | - | 1 | 809 | |||