siRNA Details
| Gene Symbol: Samd11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110516.1 | NP_001103986.1 | 11 | 1993 | ||
| GenBank mRNA | AK149268.1 | - | 11 | 1993 | ||
| AK161035.1 | - | 10 | 2007 | |||
| AY458844.1 | - | 10 | 1810 | |||
| BC026511.1 | - | 6 | 1881 | |||
| BC026991.1 | - | 10 | 2115 | 5363470 | ||

Samd11
Genomic Map