siRNA Details
| Gene Symbol: Prr11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175563.5 | NP_780772.1 | 10 | 2020 | ||
| GenBank mRNA | AK083651.1 | - | 10 | 2015 | ||
| BC023918.1 | - | 10 | 2030 | 5323297 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prr11| Gene Symbol: Prr11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175563.5 | NP_780772.1 | 10 | 2020 | ||
| GenBank mRNA | AK083651.1 | - | 10 | 2015 | ||
| BC023918.1 | - | 10 | 2030 | 5323297 | ||