siRNA Details
| Gene Symbol: Hist1h3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175653.2 | NP_783584.1 | 1 | 432 | ||
| GenBank mRNA | BC120800.1 | - | 1 | 513 | 40129723 | |
| BC120802.1 | - | 1 | 513 | 40129725 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hist1h3c| Gene Symbol: Hist1h3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175653.2 | NP_783584.1 | 1 | 432 | ||
| GenBank mRNA | BC120800.1 | - | 1 | 513 | 40129723 | |
| BC120802.1 | - | 1 | 513 | 40129725 | ||