siRNA Details
Gene Symbol: D030056L22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177640.4 | NP_808308.1 | 1, 2 | 428 | ||
GenBank mRNA | AK083631.1 | - | 1, 2 | 429 | ||
AK155389.1 | - | 1, 2 | 469 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: D030056L22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177640.4 | NP_808308.1 | 1, 2 | 428 | ||
GenBank mRNA | AK083631.1 | - | 1, 2 | 429 | ||
AK155389.1 | - | 1, 2 | 469 |