siRNA Details
| Gene Symbol: Dolk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177648.3 | NP_808316.1 | 1 | 986 | ||
| GenBank mRNA | AB073968.1 | - | 1 | 929 | ||
| AK077581.1 | - | 1 | 956 | |||
| AK144705.1 | - | 1 | 967 | |||
| BC019543.1 | - | 1 | 931 | |||
| BC026973.1 | - | 1 | 711 | 5371267 | ||

Dolk
Genomic Map