siRNA Details
| Gene Symbol: AI467606 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178901.3 | NP_849232.1 | 2 | 1887 | ||
| GenBank mRNA | AK052739.1 | - | 2 | 1888 | ||
| AK155213.1 | - | 2 | 1943 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AI467606| Gene Symbol: AI467606 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178901.3 | NP_849232.1 | 2 | 1887 | ||
| GenBank mRNA | AK052739.1 | - | 2 | 1888 | ||
| AK155213.1 | - | 2 | 1943 | |||