siRNA Details
| Gene Symbol: 4930506M07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114312.1 | NP_001107784.1 | 17 | 4071 | ||
| NM_175172.4 | NP_780381.1 | 15 | 3757 | |||
| GenBank mRNA | AK082304.1 | - | 15 | 3758 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930506M07Rik| Gene Symbol: 4930506M07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114312.1 | NP_001107784.1 | 17 | 4071 | ||
| NM_175172.4 | NP_780381.1 | 15 | 3757 | |||
| GenBank mRNA | AK082304.1 | - | 15 | 3758 | ||