siRNA Details
Gene Symbol: Klhl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178633.3 | NP_848748.2 | 3198 | |||
GenBank mRNA | AK032116.1 | - | 2746 | |||
AK079387.1 | - | 2944 | ||||
AK167903.1 | - | 2989 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Klhl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178633.3 | NP_848748.2 | 3198 | |||
GenBank mRNA | AK032116.1 | - | 2746 | |||
AK079387.1 | - | 2944 | ||||
AK167903.1 | - | 2989 |