siRNA Details
Gene Symbol: Prr11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175563.5 | NP_780772.1 | 10 | 2044 | ||
GenBank mRNA | AK083651.1 | - | 10 | 2039 | ||
BC023918.1 | - | 10 | 2054 | 5323297 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Prr11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175563.5 | NP_780772.1 | 10 | 2044 | ||
GenBank mRNA | AK083651.1 | - | 10 | 2039 | ||
BC023918.1 | - | 10 | 2054 | 5323297 |