siRNA Details
| Gene Symbol: Ctla2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145799.1 | NP_001139271.1 | 2 | 188 | ||
| NM_007796.2 | NP_031822.2 | 2 | 226 | |||
| GenBank mRNA | AA451288.1 | - | 2 | 188 | ||
| AK005471.1 | - | 2 | 163 | |||
| AK018801.1 | - | 2 | 181 | |||
| AK132290.1 | - | 2 | 159 | |||
| AK167433.1 | - | 2 | 159 | |||
| AK167664.1 | - | 2 | 160 | |||
| AY034577.1 | - | 2 | 135 | |||
| BC028437.1 | - | 2 | 152 | 1244311 | ||
| X15591.1 | - | 2 | 135 | |||

Ctla2a
Genomic Map