siRNA Details
Gene Symbol: Fxyd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008557.2 | NP_032583.1 | 5, 6 | 272 | ||
GenBank mRNA | AA959274.1 | - | 5, 6 | 229 | ||
AK145206.1 | - | 243 | ||||
AK166426.1 | - | 5, 6 | 245 | |||
BC002039.1 | - | 5, 6 | 236 | 3488180 | ||
BC051033.1 | - | 5, 6 | 230 | 4460667 | ||
BC056223.1 | - | 5, 6 | 240 | 6485767 | ||
X93038.1 | - | 5, 6 | 268 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
