siRNA Details
| Gene Symbol: Bex1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009052.2 | NP_033078.2 | 3 | 828 | ||
| GenBank mRNA | AF051347.1 | - | 3 | 761 | ||
| AF097438.1 | - | 3 | 773 | |||
| AK003429.1 | - | 1 | 443 | |||
| BC058805.1 | - | 1 | 1100 | |||
| BC061179.1 | - | 3 | 773 | 6531808 | ||
| BC089464.1 | - | 3 | 822 | 6594252 | ||
| CB194115.1 | - | 3 | 750 | |||

Bex1
Genomic Map