siRNA Details
Gene Symbol: IFIT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001547.4 | NP_001538.4 | 2 | 2475 | ||
GenBank mRNA | BC032839.2 | - | 2 | 2378 | 4838844 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: IFIT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001547.4 | NP_001538.4 | 2 | 2475 | ||
GenBank mRNA | BC032839.2 | - | 2 | 2378 | 4838844 |