siRNA Details
| Gene Symbol: Txn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011660.3 | NP_035790.1 | 2, 3 | 326 | ||
| GenBank mRNA | AK007537.1 | - | 2, 3 | 207 | ||
| AK007790.1 | - | 2, 3 | 204 | |||
| BC010756.1 | - | 2, 3 | 181 | 4216106 | ||
| BC094415.1 | - | 2, 3 | 188 | 3376881 | ||
| X77585.1 | - | 2, 3 | 199 | |||

Txn1
Genomic Map