siRNA Details
| Gene Symbol: Cox7c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007749.3 | NP_031775.1 | 2 | 219 | ||
| GenBank mRNA | AK002494.1 | - | 2 | 186 | ||
| AK027989.1 | - | 2 | 181 | |||
| AK131871.1 | - | 2 | 215 | |||
| AK153444.1 | - | 2 | 191 | |||
| AK168077.1 | - | 2 | 207 | |||
| AV056265.1 | - | 2 | 59 | |||
| BB696274.1 | - | 2 | 229 | |||
| BC010772.1 | - | 2 | 201 | 4216583 | ||
| BC086792.1 | - | 2 | 191 | 6411273 | ||
| X52940.1 | - | 2 | 185 | |||

Cox7c
Genomic Map