siRNA Details
| Gene Symbol: Sprr2j-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019074.1 | - | 2 | 276 | ||
| AY158994.1 | - | 2 | 279 | |||
| BC125542.1 | - | 2 | 250 | |||
| BC125544.1 | - | 2 | 250 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sprr2j-ps| Gene Symbol: Sprr2j-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019074.1 | - | 2 | 276 | ||
| AY158994.1 | - | 2 | 279 | |||
| BC125542.1 | - | 2 | 250 | |||
| BC125544.1 | - | 2 | 250 | |||