siRNA Details
| Gene Symbol: ING2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291959.1 | NP_001278888.1 | 2 | 921 | ||
| NM_001564.3 | NP_001555.1 | 2 | 1002 | |||
| XM_011531927.2 | XP_011530229.1 | 2 | 847 | |||
| GenBank mRNA | AB012853.1 | - | 2 | 947 | ||
| AF053537.1 | - | 2 | 923 | |||
| AK294310.1 | - | 2 | 943 | |||
| BC030128.2 | - | 2 | 982 | 3941655 | ||

ING2
Genomic Map