siRNA Details
| Gene Symbol: Selenbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019414.2 | NP_062287.1 | 5 | 484 | ||
| GenBank mRNA | BC024106.1 | - | 5 | 482 | 5096831 | |
| S56599.1 | - | 5 | 484 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Selenbp2| Gene Symbol: Selenbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019414.2 | NP_062287.1 | 5 | 484 | ||
| GenBank mRNA | BC024106.1 | - | 5 | 482 | 5096831 | |
| S56599.1 | - | 5 | 484 | |||