siRNA Details
| Gene Symbol: Cited4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019563.2 | NP_062509.1 | 1 | 734 | ||
| GenBank mRNA | AF143369.1 | - | 1 | 704 | ||
| AK012136.1 | - | 1 | 726 | |||
| AK154030.1 | - | 1 | 725 | |||
| BC025116.1 | - | 2 | 406 | |||
| BC080723.1 | - | 2 | 491 | |||
| BC132360.1 | - | 1 | 681 | 40130637 | ||
| BC132362.1 | - | 1 | 681 | 40130639 | ||

Cited4
Genomic Map