siRNA Details
| Gene Symbol: Foxo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019740.2 | NP_062714.1 | 3 | 1020 | ||
| GenBank mRNA | AF114259.1 | - | 3 | 1020 | ||
| AK047413.1 | - | 3 | 978 | |||
| AK139629.1 | - | 3 | 1006 | |||
| BC019532.1 | - | 2 | 860 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001106395.

Foxo3
Genomic Map