siRNA Details
| Gene Symbol: Olfr544 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020289.2 | NP_064685.2 | 2 | 784 | ||
| GenBank mRNA | AF121982.1 | - | 1 | 216 | ||
| BC106830.2 | - | 1 | 608 | 40044631 | ||
| BC126870.1 | - | 1 | 608 | 40044632 | ||
| BC126871.1 | - | 1 | 608 | 40044633 | ||

Olfr544
Genomic Map