siRNA Details
Gene Symbol: Dtymk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105667.1 | NP_001099137.1 | 2 | 249 | ||
NM_023136.2 | NP_075625.1 | 2 | 249 | |||
GenBank mRNA | AA014471.1 | - | 1 | 15 | ||
AK009220.1 | - | 2 | 222 | |||
AK011955.1 | - | 2 | 192 | |||
AK019284.1 | - | 2 | 184 | |||
AK019288.1 | - | 2 | 223 | |||
AK019445.1 | - | 2 | 219 | |||
AK088416.1 | - | 2 | 209 | |||
AK160747.1 | - | 2 | 219 | |||
AK210481.1 | - | 2 | 59 | |||
BC030178.1 | - | 2 | 211 | 5039765 | ||
BC069902.1 | - | 2 | 220 | 5026527 | ||
BY098372.1 | - | 2 | 250 | |||
CB182101.1 | - | 2 | 205 | |||
CF983443.1 | - | 2 | 237 | |||
S82852.1 | - | 2 | 190 | |||
S82853.1 | - | 2 | 190 |
