siRNA Details
| Gene Symbol: Uba7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023738.4 | NP_076227.1 | 2 | 196 | ||
| GenBank mRNA | AK004894.2 | - | 2 | 197 | ||
| AK054410.1 | - | 2 | 203 | |||
| AK089235.1 | - | 2 | 200 | |||
| AK089256.1 | - | 2 | 182 | |||
| AK129009.1 | - | 2 | 108 | |||
| AK170421.1 | - | 2 | 189 | |||
| AK170644.1 | - | 2 | 195 | |||
| AK172467.1 | - | 2 | 158 | |||
| BC138843.1 | - | 2 | 113 | 8861865 | ||

Uba7
Genomic Map