siRNA Details
| Gene Symbol: Akr1c12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013777.2 | NP_038805.2 | 3 | 339 | ||
| GenBank mRNA | AB027237.1 | - | 3 | 309 | ||
| AF177041.1 | - | 3 | 330 | |||
| AK007716.1 | - | 3 | 337 | |||
| BC012643.1 | - | 3 | 313 | 4205715 | ||
| BC063780.1 | - | 3 | 316 | 4219625 | ||

Akr1c12
Genomic Map