siRNA Details
| Gene Symbol: Il17b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019508.1 | NP_062381.1 | 2 | 219 | ||
| GenBank mRNA | AF184970.1 | - | 2 | 219 | ||
| AF218726.1 | - | 2 | 219 | |||
| AF250308.1 | - | 2 | 216 | |||
| AK003506.1 | - | 2 | 277 | |||
| BC002271.1 | - | 2 | 219 | 3496106 | ||

Il17b
Genomic Map