siRNA Details
| Gene Symbol: Crybb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159650.1 | NP_001153122.1 | 3 | 153 | ||
| NM_021352.3 | NP_067327.1 | 3 | 223 | |||
| GenBank mRNA | AJ272229.2 | - | 2 | 207 | ||
| AK053552.1 | - | 1 | 537 | |||
| BC031442.1 | - | 2 | 214 | 4501690 | ||
| BC058522.1 | - | 2 | 132 | 6824229 | ||
| CJ046471.1 | - | 3 | 224 | |||

Crybb3
Genomic Map