siRNA Details
| Gene Symbol: Krtap3-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025524.2 | NP_079800.1 | 1 | 388 | ||
| GenBank mRNA | AK009052.1 | - | 1 | 392 | ||
| AK009410.1 | - | 1 | 390 | |||
| AK010142.1 | - | 1 | 389 | |||
| BC120760.1 | - | 1 | 356 | 40129683 | ||
| BC120762.1 | - | 1 | 356 | 40129685 | ||

Krtap3-3
Genomic Map