siRNA Details
Gene Symbol: Tmem107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025838.2 | NP_080114.1 | 2 | 238 | ||
NM_028336.3 | NP_082612.2 | 2 | 238 | |||
GenBank mRNA | AK002804.1 | - | 2 | 208 | ||
AK003403.1 | - | 2 | 234 | |||
AK012938.1 | - | 2 | 237 | |||
AK154279.1 | - | 2 | 236 | |||
BC096686.1 | - | 2 | 216 | 1446796 | ||
BC120499.1 | - | 2 | 160 | 40129422 | ||
BC137584.1 | - | 2 | 161 | 8860603 |

More Information
Other Species Targeted

Gene Ontology Categories:
