siRNA Details
| Gene Symbol: Slc19a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030556.2 | NP_085033.2 | 4 | 580 | ||
| GenBank mRNA | AF271634.1 | - | 4 | 580 | ||
| BC109154.1 | - | 4 | 563 | 40047017 | ||
| BC109155.1 | - | 3 | 433 | 40047019 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc19a3| Gene Symbol: Slc19a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030556.2 | NP_085033.2 | 4 | 580 | ||
| GenBank mRNA | AF271634.1 | - | 4 | 580 | ||
| BC109154.1 | - | 4 | 563 | 40047017 | ||
| BC109155.1 | - | 3 | 433 | 40047019 | ||