siRNA Details
| Gene Symbol: Prrt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030890.1 | NP_112152.1 | 4 | 777 | ||
| GenBank mRNA | BC115512.2 | - | 4 | 580 | ||
| BC115513.2 | - | 4 | 580 | |||
| BC128489.1 | - | 5 | 446 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prrt1| Gene Symbol: Prrt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030890.1 | NP_112152.1 | 4 | 777 | ||
| GenBank mRNA | BC115512.2 | - | 4 | 580 | ||
| BC115513.2 | - | 4 | 580 | |||
| BC128489.1 | - | 5 | 446 | |||