siRNA Details
| Gene Symbol: Klra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF419250.1 | - | 6 | 996 | ||
| AF444273.1 | - | 6 | 996 | |||
| Gene Symbol: Klra22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053152.2 | NP_444382.1 | 4, 5 | 657 | ||
| GenBank mRNA | AF288381.1 | - | 4, 5 | 657 | ||

Klra1
Genomic Map