siRNA Details
| Gene Symbol: Klra19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053154.2 | NP_444384.1 | 2, 3 | 222 | ||
| GenBank mRNA | AF288378.1 | - | 2, 3 | 222 | ||
| BC116822.1 | - | 2, 3 | 181 | 40126141 | ||
| BC116824.1 | - | 2, 3 | 175 | 40126143 | ||

Klra19
Genomic Map