siRNA Details
| Gene Symbol: Defb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054074.1 | NP_473415.1 | 2 | 115 | ||
| GenBank mRNA | AB063109.1 | - | 2 | 115 | ||
| BC119422.1 | - | 2 | 115 | 40038954 | ||
| BC119423.2 | - | 2 | 115 | 40038955 | ||
| BC141871.1 | - | 2 | 115 | 40038967 | ||
| BC141872.1 | - | 2 | 115 | 40038968 | ||

Defb6
Genomic Map