siRNA Details
| Gene Symbol: AMPH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001635.3 | NP_001626.1 | 21 | 2878 | ||
| NM_139316.2 | NP_647477.1 | 20 | 2752 | |||
| XM_006715689.3 | XP_006715752.1 | 20 | 4415 | |||
| XM_006715690.3 | XP_006715753.1 | 20 | 3197 | |||
| XM_011515271.2 | XP_011513573.1 | 21 | 3323 | |||
| XM_017011995.1 | XP_016867484.1 | 21 | 4541 | |||
| XM_017011996.1 | XP_016867485.1 | 20 | 3503 | |||
| GenBank mRNA | AF034996.1 | - | 20 | 2605 | ||
| BC034376.1 | - | 21 | 2669 | 5181100 | ||
| X81438.1 | - | 21 | 2736 | |||

AMPH
Genomic Map