siRNA Details
Gene Symbol: Olfr74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054091.2 | NP_473432.2 | 1 | 84 | ||
GenBank mRNA | AB061229.1 | - | 1 | 84 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054091.2 | NP_473432.2 | 1 | 84 | ||
GenBank mRNA | AB061229.1 | - | 1 | 84 |