siRNA Details
| Gene Symbol: Yipf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026417.4 | NP_080693.2 | 2 | 331 | ||
| GenBank mRNA | AK009618.1 | - | 2 | 317 | ||
| AK017721.1 | - | 2 | 309 | |||
| AK017722.1 | - | 2 | 309 | |||
| AK083301.1 | - | 2 | 311 | |||
| AK159227.1 | - | 2 | 292 | |||
| AK166347.1 | - | 2 | 319 | |||
| BC003936.1 | - | 3 | 506 | 3486503 | ||

Yipf4
Genomic Map