siRNA Details
| Gene Symbol: Ifitm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001112715.1 | NP_001106186.1 | 2 | 448 | ||
| NM_026820.3 | NP_081096.3 | 3 | 422 | |||
| GenBank mRNA | AK004121.1 | - | 2 | 311 | ||
| AK169960.1 | - | 2 | 293 | |||
| BC027285.1 | - | 3 | 638 | 3492212 | ||
| BC090258.1 | - | 2 | 276 | 4982747 | ||
| BC090972.1 | - | 2 | 376 | 30297959 | ||

Ifitm1
Genomic Map