siRNA Details
| Gene Symbol: 2310057N15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027170.1 | NP_081446.1 | 1 | 537 | ||
| GenBank mRNA | AK009968.1 | - | 1 | 537 | ||
| BC104340.1 | - | 1 | 479 | |||
| BC104341.1 | - | 1 | 481 | 40049406 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2310057N15Rik| Gene Symbol: 2310057N15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027170.1 | NP_081446.1 | 1 | 537 | ||
| GenBank mRNA | AK009968.1 | - | 1 | 537 | ||
| BC104340.1 | - | 1 | 479 | |||
| BC104341.1 | - | 1 | 481 | 40049406 | ||