siRNA Details
| Gene Symbol: Ankrd33b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164441.1 | NP_001157913.1 | 1 | 499 | ||
| NM_026153.3 | NP_080429.1 | 1 | 499 | |||
| NM_027496.3 | NP_081772.1 | 1 | 499 | |||
| GenBank mRNA | AK002572.1 | - | 1 | 294 | ||
| AK012871.1 | - | 1 | 307 | |||
| AK017844.1 | - | 1 | 304 | |||
| AK089667.1 | - | 1 | 273 | |||
| AK156757.1 | - | 1 | 467 | |||
| AK170894.1 | - | 1 | 273 | |||
| BC100327.1 | - | 1 | 499 | |||

Ankrd33b
Genomic Map