siRNA Details
| Gene Symbol: 2310067B10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028014.3 | NP_082290.2 | 32 | 4674 | ||
| GenBank mRNA | AK010082.1 | - | 3 | 673 | ||
| AK047622.1 | - | 18 | 2808 | |||
| BC007157.1 | - | 19 | 2920 | |||
| BC030865.1 | - | 17 | 2441 | |||
| BC053088.1 | - | 32 | 4589 | 5709104 | ||
| CA325552.1 | - | 1 | 171 | |||

2310067B10Rik
Genomic Map