siRNA Details
| Gene Symbol: Vmn1r25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053238.2 | NP_444468.2 | 1 | 803 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r25| Gene Symbol: Vmn1r25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053238.2 | NP_444468.2 | 1 | 803 | ||