siRNA Details
| Gene Symbol: Fbxl15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133694.2 | NP_598455.2 | 3 | 888 | ||
| GenBank mRNA | AK003032.1 | - | 3 | 611 | ||
| AK145009.1 | - | 3 | 848 | |||
| AK172339.1 | - | 4 | 785 | |||
| AW825726.1 | - | 1 | 294 | |||
| BC016499.1 | - | 3 | 863 | 4221584 | ||
| CK791452.1 | - | 3 | 574 | |||

Fbxl15
Genomic Map