siRNA Details
| Gene Symbol: Vmn1r6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134175.1 | NP_598936.1 | 1 | 325 | ||
| GenBank mRNA | BC125532.1 | - | 1 | 380 | 40130047 | |
| BC125534.1 | - | 1 | 380 | 40130049 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r6| Gene Symbol: Vmn1r6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134175.1 | NP_598936.1 | 1 | 325 | ||
| GenBank mRNA | BC125532.1 | - | 1 | 380 | 40130047 | |
| BC125534.1 | - | 1 | 380 | 40130049 | ||