siRNA Details
| Gene Symbol: Olfr981 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146286.1 | NP_666398.1 | 1 | 248 | ||
| GenBank mRNA | BC120776.1 | - | 1 | 291 | 40129699 | |
| BC120778.1 | - | 1 | 291 | 40129701 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr981| Gene Symbol: Olfr981 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146286.1 | NP_666398.1 | 1 | 248 | ||
| GenBank mRNA | BC120776.1 | - | 1 | 291 | 40129699 | |
| BC120778.1 | - | 1 | 291 | 40129701 | ||